Assay Details
Gene Symbol: LZIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009000383.2 | XP_008998631.1 | 6 - 7 | 780 | 61 | |
XM_009000384.2 | XP_008998632.1 | 6 - 7 | 689 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
