Assay Details
| Gene Symbol: TMEM205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761754.4 | XP_002761800.1 | 3 - 4 | 650 | 65 | |
| XM_008987298.2 | XP_008985546.1 | 3 - 4 | 379 | 65 | ||
| XM_008987299.2 | XP_008985547.1 | 3 - 4 | 370 | 65 | ||
More Information
Set Membership: |
|

TMEM205
Genomic Map