Assay Details
Gene Symbol: PEMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002747882.3 | XP_002747928.1 | 2 - 3 | 366 | 61 | |
XM_008996455.2 | XP_008994703.1 | 2 - 3 | 312 | 61 | ||
XM_008996457.2 | XP_008994705.1 | 2 - 3 | 129 | 61 | ||
XM_008996458.1 | XP_008994706.1 | 2 - 3 | 172 | 61 | ||
XM_008996460.2 | XP_008994708.1 | 2 - 3 | 275 | 61 | ||
XM_008996461.2 | XP_008994709.1 | 2 - 3 | 198 | 61 | ||
XM_017972250.1 | XP_017827739.1 | 2 - 3 | 131 | 61 | ||
XM_017972251.1 | XP_017827740.1 | 2 - 3 | 308 | 61 | ||
XM_017972252.1 | XP_017827741.1 | 3 - 4 | 395 | 61 | ||
XM_017972253.1 | XP_017827742.1 | 3 - 4 | 384 | 61 | ||
XM_017972254.1 | XP_017827743.1 | 2 - 3 | 131 | 61 | ||
XM_017972255.1 | XP_017827744.1 | 2 - 3 | 204 | 61 | ||
XM_017972256.1 | XP_017827745.1 | 1 - 2 | 120 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
