Assay Details
| Gene Symbol: MOGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749832.3 | XP_002749878.1 | 5 - 6 | 954 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MOGAT1| Gene Symbol: MOGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749832.3 | XP_002749878.1 | 5 - 6 | 954 | 78 | |
Set Membership: |
|