Assay Details
Gene Symbol: CERS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002752447.2 | XP_002752493.1 | 1 - 2 | 432 | 63 | |
XM_009003767.1 | XP_009002015.1 | 1 - 2 | 432 | 63 | ||
XM_017976047.1 | XP_017831536.1 | 1 - 2 | 432 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
