Assay Details
Gene Symbol: LRRC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002761170.3 | XP_002761216.1 | 9 - 10 | 1546 | 60 | |
XM_008986049.2 | XP_008984297.1 | 9 - 10 | 1546 | 60 | ||
XM_008986050.2 | XP_008984298.1 | 9 - 10 | 1546 | 60 | ||
XM_008986052.2 | XP_008984300.1 | 9 - 10 | 1192 | 60 | ||
XM_008986053.2 | XP_008984301.1 | 8 - 9 | 1105 | 60 | ||
XM_008986054.2 | XP_008984302.1 | 9 - 10 | 1546 | 60 | ||
XM_008986055.2 | XP_008984303.1 | 9 - 10 | 1546 | 60 | ||
XM_008986056.2 | XP_008984304.1 | 9 - 10 | 1520 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
