Assay Details
Gene Symbol: RBBP8NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002747745.2 | XP_002747791.2 | 353 | 74 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
