Assay Details
| Gene Symbol: BRMS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002807371.4 | XP_002807417.2 | 2 - 3 | 293 | 61 | |
| XM_009008511.2 | XP_009006759.1 | 2 - 3 | 296 | 61 | ||
| XM_017978294.1 | XP_017833783.1 | 2 - 3 | 298 | 61 | ||
More Information
Set Membership: |
|

BRMS1
Genomic Map