Assay Details
| Gene Symbol: SLC16A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756340.3 | XP_002756386.1 | 4 - 5 | 884 | 88 | |
| XM_009009897.2 | XP_009008145.1 | 4 - 5 | 491 | 88 | ||
| XM_017979097.1 | XP_017834586.1 | 2 - 3 | 323 | 88 | ||
More Information
Set Membership: |
|

SLC16A9
Genomic Map