Assay Details
Gene Symbol: MEPCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744001.4 | XP_002744047.1 | 1 - 2 | 2594 | 75 | |
XM_008982605.1 | XP_008980853.1 | 2 - 3 | 906 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
