Assay Details
Gene Symbol: RLIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002763014.4 | XP_002763060.1 | 3 - 4 | 476 | 98 | |
XM_008989510.2 | XP_008987758.1 | 4 - 5 | 540 | 98 | ||
XM_017968560.1 | XP_017824049.1 | 4 - 5 | 362 | 98 | ||
XM_017968561.1 | XP_017824050.1 | 3 - 4 | 335 | 98 | ||
XM_017968562.1 | XP_017824051.1 | 3 - 4 | 1101 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
