Assay Details
Gene Symbol: CDC42SE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744625.2 | XP_002744671.1 | 3 - 4 | 562 | 86 | |
XM_008991548.1 | XP_008989796.1 | 4 - 5 | 794 | 86 | ||
XM_008991555.1 | XP_008989803.1 | 4 - 5 | 794 | 86 | ||
XM_008991558.1 | XP_008989806.1 | 4 - 5 | 794 | 86 | ||
XM_008991559.1 | XP_008989807.1 | 3 - 4 | 600 | 86 | ||
XM_017969703.1 | XP_017825192.1 | 4 - 5 | 794 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
