Assay Details
| Gene Symbol: CCDC129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733566.3 | XP_003733614.3 | 12 - 13 | 3102 | 79 | |
| XM_017975216.1 | XP_017830705.1 | 12 - 13 | 2839 | 79 | ||
More Information
Set Membership: |
|

CCDC129
Genomic Map