Assay Details
| Gene Symbol: P2RY13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759488.3 | XP_002759534.3 | 1 - 2 | 3468 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

P2RY13| Gene Symbol: P2RY13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759488.3 | XP_002759534.3 | 1 - 2 | 3468 | 67 | |
Set Membership: |
|