Assay Details
| Gene Symbol: TNFRSF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017974334.1 | XP_017829823.1 | 7 - 8 | 976 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TNFRSF1B| Gene Symbol: TNFRSF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017974334.1 | XP_017829823.1 | 7 - 8 | 976 | 74 | |
Set Membership: |
|