Assay Details
Gene Symbol: BAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002755473.3 | XP_002755519.1 | 2 - 3 | 247 | 116 | |
XM_009008400.2 | XP_009006648.1 | 2 - 3 | 673 | 116 | ||
XM_009008401.2 | XP_009006649.1 | 2 - 3 | 307 | 116 | ||
XM_009008402.2 | XP_009006650.1 | 2 - 3 | 217 | 116 | ||
XM_017978230.1 | XP_017833719.1 | 2 - 3 | 738 | 116 |

More Information
Set Membership: |
![]() ![]() |
