Assay Details
| Gene Symbol: B2M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753411.4 | XP_002753457.2 | 3 - 4 | 424 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

B2M| Gene Symbol: B2M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753411.4 | XP_002753457.2 | 3 - 4 | 424 | 75 | |
Set Membership: |
|