Assay Details
Gene Symbol: C21H3orf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003735367.3 | XP_003735415.1 | 1 - 2 | 460 | 94 | |
XM_008986452.2 | XP_008984700.1 | 1 - 2 | 460 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
