Assay Details
Gene Symbol: HDHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002743217.4 | XP_002743263.1 | 2 | 898 | 83 | |
XM_009001595.2 | XP_008999843.1 | 2 | 886 | 83 | ||
XM_009001605.2 | XP_008999853.1 | 2 | 943 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
