Assay Details
| Gene Symbol: VMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748280.3 | XP_002748326.1 | 2 - 3 | 984 | 59 | |
| XM_008997235.2 | XP_008995483.1 | 2 - 3 | 995 | 59 | ||
More Information
Set Membership: |
|

VMO1
Genomic Map