Assay Details
Gene Symbol: RAB11FIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008980512.2 | XP_008978760.1 | 1 - 2 | 629 | 79 | |
XM_008980513.2 | XP_008978761.1 | 1 - 2 | 629 | 79 | ||
XM_008980515.2 | XP_008978763.1 | 1 - 2 | 629 | 79 | ||
XM_017964243.1 | XP_017819732.1 | 1 - 2 | 629 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
