Assay Details
| Gene Symbol: TFDP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759529.4 | XP_002759575.3 | 3 - 4 | 463 | 105 | |
| XM_008983727.2 | XP_008981975.1 | 3 - 4 | 463 | 105 | ||
| XM_008983728.2 | XP_008981976.1 | 3 - 4 | 261 | 105 | ||
| XM_008983731.2 | XP_008981979.1 | 3 - 4 | 261 | 105 | ||
| XM_017965784.1 | XP_017821273.1 | 2 - 3 | 133 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008983728.2, XM_008983731.2, XM_017965784.1 transcripts.
Set Membership: |
|

TFDP2
Genomic Map