Assay Details
Gene Symbol: CRADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002752856.3 | XP_002752902.1 | 2 - 3 | 432 | 77 | |
XM_017976281.1 | XP_017831770.1 | 2 - 3 | 421 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
