Assay Details
| Gene Symbol: TMEM184B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743771.4 | XP_002743817.1 | 3 - 4 | 566 | 90 | |
| XM_008979089.2 | XP_008977337.1 | 3 - 4 | 566 | 90 | ||
| XM_008979095.2 | XP_008977343.1 | 3 - 4 | 631 | 90 | ||
| XM_008979106.2 | XP_008977354.1 | 4 - 5 | 643 | 90 | ||
| XM_017963679.1 | XP_017819168.1 | 2 - 3 | 1559 | 90 | ||
More Information
Set Membership: |
|

TMEM184B
Genomic Map