Assay Details
Gene Symbol: CGREF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003734771.3 | XP_003734819.1 | 2 - 3 | 376 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
