Assay Details
| Gene Symbol: GUCY1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009006719.2 | XP_009004967.2 | 5 - 6 | 1134 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUCY1A2| Gene Symbol: GUCY1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009006719.2 | XP_009004967.2 | 5 - 6 | 1134 | 69 | |
Set Membership: |
|