Assay Details
Gene Symbol: SPACA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008988521.2 | XP_008986769.1 | 3 - 4 | 2535 | 65 | |
XM_017968064.1 | XP_017823553.1 | 3 - 4 | 239 | 65 | ||
XM_017968065.1 | XP_017823554.1 | 3 - 4 | 291 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
