Assay Details
| Gene Symbol: CYSLTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017971403.1 | XP_017826892.1 | 3 | 2222 | 94 | |
| XM_017971405.1 | XP_017826894.1 | 3 | 1579 | 94 | ||
More Information
Set Membership: |
|

CYSLTR2
Genomic Map