Assay Details
| Gene Symbol: L3MBTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008995954.2 | XP_008994202.1 | 2 - 3 | 184 | 111 | |
| XM_017972060.1 | XP_017827549.1 | 1 - 2 | 199 | 111 | ||
More Information
Set Membership: |
|

L3MBTL1
Genomic Map