Assay Details
Gene Symbol: NECTIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008982497.2 | XP_008980745.1 | 1 - 2 | 422 | 64 | |
XM_008982499.2 | XP_008980747.1 | 1 - 2 | 422 | 64 | ||
XM_008982500.2 | XP_008980748.1 | 1 - 2 | 422 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
