Assay Details
Gene Symbol: LOC100392681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017973898.1 | XP_017829387.1 | 2 - 3 | 753 | 65 | |
XM_017973899.1 | XP_017829388.1 | 2 - 3 | 753 | 65 | ||
XM_017973901.1 | XP_017829390.1 | 2 - 3 | 753 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
