Assay Details
Gene Symbol: BAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002746436.3 | XP_002746482.1 | 4 - 5 | 1370 | 60 | |
XM_008994261.2 | XP_008992509.1 | 4 - 5 | 658 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
