Assay Details
| Gene Symbol: ERI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750734.4 | XP_002750780.2 | 6 - 7 | 968 | 67 | |
| XM_003733428.2 | XP_003733476.1 | 6 - 7 | 668 | 67 | ||
| XM_009001068.2 | XP_008999316.1 | 6 - 7 | 962 | 67 | ||
| XM_009001069.2 | XP_008999317.1 | 6 - 7 | 956 | 67 | ||
| XM_009001070.2 | XP_008999318.1 | 6 - 7 | 902 | 67 | ||
| XM_009001071.2 | XP_008999319.1 | 6 - 7 | 962 | 67 | ||
| XM_009001072.2 | XP_008999320.1 | 5 - 6 | 1150 | 67 | ||
| XM_009001073.1 | XP_008999321.1 | 5 - 6 | 630 | 67 | ||
| XM_009001074.2 | XP_008999322.1 | 4 - 5 | 1054 | 67 | ||
| XM_017974645.1 | XP_017830134.1 | 6 - 7 | 962 | 67 | ||
| XM_017974646.1 | XP_017830135.1 | 5 - 6 | 566 | 67 | ||
| XM_017974647.1 | XP_017830136.1 | 6 - 7 | 648 | 67 | ||
| XM_017974648.1 | XP_017830137.1 | 6 - 7 | 659 | 67 | ||
| XM_017974649.1 | XP_017830138.1 | 4 - 5 | 774 | 67 | ||
| XM_017974650.1 | XP_017830139.1 | 5 - 6 | 366 | 67 | ||
More Information
Set Membership: |
|

ERI3
Genomic Map