Assay Details
Gene Symbol: SEPT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009009112.2 | XP_009007360.2 | 4 - 5 | 665 | 67 | |
XM_009009113.2 | XP_009007361.1 | 4 - 5 | 607 | 67 | ||
XM_009009115.2 | XP_009007363.1 | 3 - 4 | 367 | 67 | ||
XM_017978699.1 | XP_017834188.1 | 4 - 5 | 665 | 67 | ||
XM_017978700.1 | XP_017834189.1 | 4 - 5 | 665 | 67 | ||
XM_017978701.1 | XP_017834190.1 | 4 - 5 | 609 | 67 | ||
XM_017978702.1 | XP_017834191.1 | 2 - 3 | 451 | 67 | ||
XM_017978703.1 | XP_017834192.1 | 3 - 4 | 545 | 67 | ||
XM_017978704.1 | XP_017834193.1 | 5 - 6 | 503 | 67 | ||
XM_017978705.1 | XP_017834194.1 | 4 - 5 | 409 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
