Assay Details
| Gene Symbol: TRAPPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750614.2 | XP_002750660.1 | 4 - 5 | 564 | 114 | |
| XM_009000894.1 | XP_008999142.1 | 4 - 5 | 825 | 114 | ||
| XM_009000895.1 | XP_008999143.1 | 2 - 3 | 366 | 114 | ||
More Information
Set Membership: |
|

TRAPPC3
Genomic Map