Assay Details
Gene Symbol: ZNF789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744029.3 | XP_002744075.1 | 4 - 5 | 543 | 74 | |
XM_008983069.2 | XP_008981317.1 | 4 - 5 | 513 | 74 | ||
XM_008983096.2 | XP_008981344.1 | 3 - 4 | 371 | 74 | ||
XM_017966590.1 | XP_017822079.1 | 3 - 4 | 702 | 74 | ||
XM_017966594.1 | XP_017822083.1 | 3 - 4 | 683 | 74 | ||
XM_017966599.1 | XP_017822088.1 | 4 - 5 | 1153 | 74 | ||
XM_017966600.1 | XP_017822089.1 | 4 - 5 | 429 | 74 | ||
XM_017966608.1 | XP_017822097.1 | 2 - 3 | 389 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
