Assay Details
| Gene Symbol: VSIG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754546.3 | XP_002754592.1 | 6 - 7 | 1764 | 83 | |
| XM_009007040.2 | XP_009005288.1 | 5 - 6 | 1237 | 83 | ||
More Information
Set Membership: |
|

VSIG2
Genomic Map