Assay Details
Gene Symbol: NDUFAF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008980857.2 | XP_008979105.1 | 7 - 8 | 879 | 67 | |
XM_017964410.1 | XP_017819899.1 | 6 - 7 | 649 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
