Assay Details
| Gene Symbol: RNF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753102.4 | XP_002753148.1 | 5 - 6 | 1102 | 105 | |
| XM_017976440.1 | XP_017831929.1 | 4 - 5 | 647 | 105 | ||
More Information
Set Membership: |
|

RNF34
Genomic Map