Assay Details
| Gene Symbol: SPINT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987930.2 | XP_008986178.1 | 4 - 5 | 863 | 65 | |
| XM_008987931.2 | XP_008986179.1 | 4 - 5 | 863 | 65 | ||
| XM_008987932.2 | XP_008986180.1 | 3 - 4 | 803 | 65 | ||
| XM_008987934.2 | XP_008986182.1 | 3 - 4 | 803 | 65 | ||
| XM_008987937.2 | XP_008986185.1 | 3 - 4 | 692 | 65 | ||
| XM_008987938.2 | XP_008986186.1 | 3 - 4 | 692 | 65 | ||
More Information
Set Membership: |
|

SPINT2
Genomic Map