Assay Details
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009006452.2 | XP_009004700.1 | 2 - 3 | 777 | 98 | |
XM_009006464.2 | XP_009004712.1 | 2 - 3 | 267 | 98 | ||
XM_017977820.1 | XP_017833309.1 | 2 - 3 | 175 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009006464.2, XM_017977820.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
