Assay Details
Gene Symbol: CA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002762329.3 | XP_002762375.2 | 8 - 9 | 1768 | 58 | |
GenBank mRNA | EF215379.1 | - | 5 - 6 | 599 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
