Assay Details
Gene Symbol: DHPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003735492.3 | XP_003735540.1 | 1 - 2 | 382 | 77 | |
XM_008987359.2 | XP_008985607.1 | 1 - 2 | 388 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
