Assay Details
Gene Symbol: AURKAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009000225.2 | XP_008998473.1 | 3 - 4 | 1083 | 55 | |
XM_009000226.2 | XP_008998474.1 | 3 - 4 | 844 | 55 | ||
XM_017974228.1 | XP_017829717.1 | 2 - 3 | 539 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
