Assay Details
| Gene Symbol: KIRREL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760125.3 | XP_002760171.1 | 6 - 7 | 1165 | 99 | |
| XM_008984666.2 | XP_008982914.1 | 6 - 7 | 1165 | 99 | ||
More Information
Set Membership: |
|

KIRREL1
Genomic Map