Assay Details
| Gene Symbol: FCGR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003735165.3 | XP_003735213.1 | 6 - 7 | 840 | 79 | |
| XM_008984785.2 | XP_008983033.1 | 6 - 7 | 842 | 79 | ||
| XM_008984786.2 | XP_008983034.1 | 6 - 7 | 853 | 79 | ||
More Information
Set Membership: |
|

FCGR2A
Genomic Map