Assay Details
| Gene Symbol: OCEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761874.3 | XP_002761920.1 | 5 - 6 | 718 | 65 | |
| XM_008987483.2 | XP_008985731.1 | 4 - 5 | 900 | 65 | ||
| XM_008987484.2 | XP_008985732.1 | 4 - 5 | 868 | 65 | ||
| XM_017967602.1 | XP_017823091.1 | 5 - 6 | 751 | 65 | ||
More Information
Set Membership: |
|

OCEL1
Genomic Map