Assay Details
| Gene Symbol: FMO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008985038.2 | XP_008983286.1 | 9 - 10 | 1652 | 76 | |
| XM_017966092.1 | XP_017821581.1 | 9 - 10 | 1616 | 76 | ||
| XM_017966093.1 | XP_017821582.1 | 6 - 7 | 1298 | 76 | ||
More Information
Set Membership: |
|

FMO4
Genomic Map