Assay Details
Gene Symbol: CENPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008994554.1 | XP_008992802.1 | 6 - 7 | 607 | 105 | |
XM_008994555.2 | XP_008992803.1 | 6 - 7 | 601 | 105 | ||
XM_008994556.1 | XP_008992804.1 | 5 - 6 | 487 | 105 | ||
XM_008994557.1 | XP_008992805.1 | 5 - 6 | 481 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
