Assay Details
| Gene Symbol: POLR2E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986886.2 | XP_008985134.2 | 2 - 3 | 281 | 64 | |
| XM_017967546.1 | XP_017823035.1 | 6 - 7 | 693 | 64 | ||
More Information
Set Membership: |
|

POLR2E
Genomic Map